Learn More
Medchemexpress LLC Thalidomide-O-amido-C3-PEG3-C1-NH2 | 1799711-29-7 | 99.1% | 648.58 g/mol | C27H35F3N4O11 | 5 MG
SDP
Catalog No. 5000262574
Encompass_Preferred
Thalidomide-O-amido-C3-PEG3-C1-NH2 is a thalidomide-derived cereblon E3 ligase ligand conjugated to a three-unit PEG linker and a short C3 spacer terminating in an amine. Supplied for research use, it is intended as a linker-ligand building block for PROTAC design and related chemical biology applications.
  • Thalidomide-based cereblon ligand for E3 ligase recruitment.
  • Three-unit PEG linker and C3 spacer terminating in an amine for conjugation.
  • High purity suitable for research applications (~99.1%).
  • Available in small pack sizes for discovery work (mg scale).
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • For research use only, not for human use.
Catalog No. 50-002-62574 Supplier Medchemexpress LLC Supplier No. HY1316465MG
Thalidomide-O-amido-C3-PEG3-C1-NH2 is a thalidomide-derived cereblon E3 ligase ligand conjugated to a three-unit PEG linker and a short C3 spacer terminating in an amine. Supplied for research use, it is intended as a linker-ligand building block for PROTAC design and related chemical biology applications.
  • Thalidomide-based cereblon ligand for E3 ligase recruitment.
  • Three-unit PEG linker and C3 spacer terminating in an amine for conjugation.
  • High purity suitable for research applications (~99.1%).
  • Available in small pack sizes for discovery work (mg scale).
  • Store sealed at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • For research use only, not for human use.