Learn More
TARGETMOL CHEMICALS INC Thalidomide-O-amido-C4-N3 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1518910MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes.Thalidomide-O-amido-C4-N3 (Cereblon Ligand-Linker Conjugates 4) is an E3 ligase ligand-linker conjugate incorporating the Thalidomide-based cereblon ligand and a linker. Purity 97.01%
Catalog No. 50-287-8440