Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-o-amido-c4-n3 | 2098488-36-7 | MFCD31560486 | ≥98.0% | 428.40 g·mol⁻¹ | C19H20N6O6 | 5 MG
SDP

Catalog No. 5000245071
Encompass_Preferred

Thalidomide-O-amido-C4-N3 is a thalidomide-derived cereblon ligand-linker conjugate that contains an azide functional group. It is supplied as a research-grade building block for the design and synthesis of proteolysis-targeting chimeras (PROTACs) and for click-chemistry bioconjugation, compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted azide-alkyne cycloaddition.

  • Contains an azide functional group for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • Suitable for PROTAC synthesis and E3 ligase recruitment studies.
  • Research-grade building block with defined molecular formula and molecular weight.
  • Forms a clear solution when dissolved in common organic solvents.
  • Recommended storage: store solid at 4°C under inert atmosphere; in solution -80°C for up to 6 months or -20°C for up to 1 month.

Catalog No. 50-002-45071 Supplier Medchemexpress LLC Supplier No. HY1036155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-C4-N3 is a thalidomide-derived cereblon ligand-linker conjugate that contains an azide functional group. It is supplied as a research-grade building block for the design and synthesis of proteolysis-targeting chimeras (PROTACs) and for click-chemistry bioconjugation, compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted azide-alkyne cycloaddition.

  • Contains an azide functional group for copper-catalyzed and strain-promoted azide-alkyne cycloadditions.
  • Suitable for PROTAC synthesis and E3 ligase recruitment studies.
  • Research-grade building block with defined molecular formula and molecular weight.
  • Forms a clear solution when dissolved in common organic solvents.
  • Recommended storage: store solid at 4°C under inert atmosphere; in solution -80°C for up to 6 months or -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.