Learn More
Medchemexpress LLC Thalidomide-O-amido-C4-NH2 hydrochloride | 2245697-86-1 | MFCD31656719 | 98.1% | 438.86 | C19H23ClN4O6 | 25 MG
SDP
Catalog No. 5000239850
Encompass_Preferred
Thalidomide-O-amido-C4-NH2 hydrochloride is a thalidomide-derived E3 ligase ligand-linker conjugate supplied as a hydrochloride salt for use as an amine intermediate in the synthesis of heterobifunctional PROTACs and related degrader molecules. It combines a cereblon-binding thalidomide scaffold with a reactive terminal amine to facilitate conjugation in medicinal chemistry workflows.
  • Provides cereblon-binding thalidomide scaffold for E3 ligase recruitment.
  • Terminal primary amine suitable for conjugation to linkers or targeting ligands.
  • High reported purity for reliable synthetic outcomes.
  • Hydrochloride salt form for improved stability and handling.
  • Available in small mg-scale packages for research synthesis.
Catalog No. 50-002-39850 Supplier Medchemexpress LLC Supplier No. HY107438A25MG
Add to Cart
Add to Cart
Thalidomide-O-amido-C4-NH2 hydrochloride is a thalidomide-derived E3 ligase ligand-linker conjugate supplied as a hydrochloride salt for use as an amine intermediate in the synthesis of heterobifunctional PROTACs and related degrader molecules. It combines a cereblon-binding thalidomide scaffold with a reactive terminal amine to facilitate conjugation in medicinal chemistry workflows.
  • Provides cereblon-binding thalidomide scaffold for E3 ligase recruitment.
  • Terminal primary amine suitable for conjugation to linkers or targeting ligands.
  • High reported purity for reliable synthetic outcomes.
  • Hydrochloride salt form for improved stability and handling.
  • Available in small mg-scale packages for research synthesis.