Learn More
Medchemexpress LLC Thalidomide-O-amido-C4-NH2 hydrochloride | 2245697-86-1 | MFCD31656719 | 98.1% | 438.86 g/mol | C19H23ClN4O6 | 100 MG
SDP
Catalog No. 5000239936
Encompass_Preferred
Thalidomide-O-amido-C4-NH2 hydrochloride is an E3 ligase (cereblon) ligand-linker intermediate supplied as the hydrochloride salt for research use. It is a white to off-white solid with documented molecular properties and high purity, intended for use in the synthesis of PROTACs and related heterobifunctional degraders.
  • High purity suitable for synthetic applications.
  • Hydrochloride salt form for improved handling and stability.
  • Solid, white to off-white appearance for easy identification.
  • Useful as a cereblon E3 ligase ligand-linker intermediate.
  • Available in small research pack sizes for laboratory use.
  • Documented molecular weight and formula for analytical reference.
Catalog No. 50-002-39936 Supplier Medchemexpress LLC Supplier No. HY107438A100MG
Thalidomide-O-amido-C4-NH2 hydrochloride is an E3 ligase (cereblon) ligand-linker intermediate supplied as the hydrochloride salt for research use. It is a white to off-white solid with documented molecular properties and high purity, intended for use in the synthesis of PROTACs and related heterobifunctional degraders.
  • High purity suitable for synthetic applications.
  • Hydrochloride salt form for improved handling and stability.
  • Solid, white to off-white appearance for easy identification.
  • Useful as a cereblon E3 ligase ligand-linker intermediate.
  • Available in small research pack sizes for laboratory use.
  • Documented molecular weight and formula for analytical reference.