Learn More
Apexbio Technology LLC Thalidomide-O-amido-C4-NH2 TFA 1799711-25-3 50mg
SDP
Catalog No. 5000093357
Encompass_Preferred
Thalidomide-O-amido-C4-NH2 TFA (CAS 1799711-25-3) is a synthetic small molecule composed of a thalidomide-derived cereblon ligand conjugated to a chemical linker This compound selectively binds to cereblon (CRBN) a substrate receptor of the CRL4CRBN E3 ubiquitin ligase complex By facilitating the recruitment of target proteins to the CRBN complex it enables their ubiquitination and subsequent proteasomal degradation Thalidomide-O-amido-C4-NH2 TFA serves as a molecular tool in the development of proteolysis targeting chimeras (PROTACs) and other targeted protein degradation strategies in biomedical research
Catalog No. 50-000-93357 Supplier Apexbio Technology LLC Supplier No. A998450
Thalidomide-O-amido-C4-NH2 TFA (CAS 1799711-25-3) is a synthetic small molecule composed of a thalidomide-derived cereblon ligand conjugated to a chemical linker This compound selectively binds to cereblon (CRBN) a substrate receptor of the CRL4CRBN E3 ubiquitin ligase complex By facilitating the recruitment of target proteins to the CRBN complex it enables their ubiquitination and subsequent proteasomal degradation Thalidomide-O-amido-C4-NH2 TFA serves as a molecular tool in the development of proteolysis targeting chimeras (PROTACs) and other targeted protein degradation strategies in biomedical research