Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-C4-NH2 (TFA) | 1799711-25-3 | MFCD31560484 | 99.9% | 516.42 g/mol | C21H23F3N4O8 | 1 G
SDP

Catalog No. 5000244962
Encompass_Preferred

Thalidomide-O-amido-C4-NH2 (TFA) is a thalidomide-derived cereblon ligand supplied as the trifluoroacetate salt and intended as a building block for E3 ligase ligand-linker conjugates and PROTAC research. The compound is provided as a solid research reagent with characterized purity, formula, and solubility data for in vitro and in vivo formulation.

  • Intended for research use only.
  • High purity 99.92%.
  • Molecular weight 516.42 g/mol.
  • Molecular formula C21H23F3N4O8.
  • Soluble in DMSO (~42.86 mg/mL); multiple in vivo formulation protocols reported.
  • Store dry at 4°C; in solvent store at -80°C for up to 6 months.

Catalog No. 50-002-44962 Supplier Medchemexpress LLC Supplier No. HY1036131G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-C4-NH2 (TFA) is a thalidomide-derived cereblon ligand supplied as the trifluoroacetate salt and intended as a building block for E3 ligase ligand-linker conjugates and PROTAC research. The compound is provided as a solid research reagent with characterized purity, formula, and solubility data for in vitro and in vivo formulation.

  • Intended for research use only.
  • High purity 99.92%.
  • Molecular weight 516.42 g/mol.
  • Molecular formula C21H23F3N4O8.
  • Soluble in DMSO (~42.86 mg/mL); multiple in vivo formulation protocols reported.
  • Store dry at 4°C; in solvent store at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.