Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-C4-NH2 (trifluoroacetate) | 1799711-25-3 | MFCD31560484 | 99.9% | 516.42 g/mol | C21H23F3N4O8 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10361350MG

Encompass_Preferred

Thalidomide-O-amido-C4-NH2 (trifluoroacetate) is a thalidomide-derived cereblon ligand supplied as the TFA salt for use as an E3 ligase ligand/linker building block in PROTAC and chemical biology research.

  • Provided as the trifluoroacetate salt.
  • High purity (99.9%, LCMS).
  • Molecular weight 516.42 g/mol.
  • CAS number 1799711-25-3.
  • Appearance: solid, white to off-white.
  • Packaged as a 50 MG quantity.

Catalog No. 50-002-46534


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.