Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-C4-NH2 trifluoroacetate | 1799711-25-3 | MFCD31560484 | 99.9% | 516.42 g/mol | C21H23F3N4O8 | 500 MG
SDP

Catalog No. 5000245014
Encompass_Preferred

Thalidomide-O-amido-C4-NH2 (TFA) is a thalidomide-derived cereblon ligand-linker conjugate supplied as the trifluoroacetate salt for use in PROTAC and degrader research. The material is provided as a white to off-white solid with documented purity, solubility, and storage recommendations to support formulation and biological testing.

  • Thalidomide-based cereblon ligand-linker conjugate (TFA salt)
  • High purity: 99.92% (LCMS)
  • Molecular weight 516.42 g/mol; formula C21H23F3N4O8
  • Solubility: DMSO 42.86 mg/mL; established in vivo vehicle protocols at ≥2.5 mg/mL
  • Storage: solid at 4°C under nitrogen; in solution -80°C (up to 6 months) or -20°C (up to 1 month)
  • Intended for use in synthesis and biological evaluation of PROTACs and E3 ligase degrader studies

Catalog No. 50-002-45014 Supplier Medchemexpress LLC Supplier No. HY103613500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-C4-NH2 (TFA) is a thalidomide-derived cereblon ligand-linker conjugate supplied as the trifluoroacetate salt for use in PROTAC and degrader research. The material is provided as a white to off-white solid with documented purity, solubility, and storage recommendations to support formulation and biological testing.

  • Thalidomide-based cereblon ligand-linker conjugate (TFA salt)
  • High purity: 99.92% (LCMS)
  • Molecular weight 516.42 g/mol; formula C21H23F3N4O8
  • Solubility: DMSO 42.86 mg/mL; established in vivo vehicle protocols at ≥2.5 mg/mL
  • Storage: solid at 4°C under nitrogen; in solution -80°C (up to 6 months) or -20°C (up to 1 month)
  • Intended for use in synthesis and biological evaluation of PROTACs and E3 ligase degrader studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.