Learn More
Medchemexpress LLC HY-P7103A 10ug Medchemexpress, IL-6 (HEK293-expressed), Rat CAS: Purity:>98%
SDP
Catalog No. 501970567
Encompass_Preferred
Medchemexpress, HY-112618A 100mg Thalidomide-O-amido-C6-NH2 (TFA) CAS:1950635-14-9 Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0567 Supplier Medchemexpress LLC Supplier No. HYP7103A10UG
Medchemexpress, HY-112618A 100mg Thalidomide-O-amido-C6-NH2 (TFA) CAS:1950635-14-9 Thalidomide-O-amido-C6-NH2 TFA (Cereblon Ligand-Linker Conjugates 11 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.