Learn More
Medchemexpress LLC Thalidomide-O-amido-C6-NH2 hydrochloride | 2376990-31-5 | 99.3% | 466.92 | 100 MG
SDP
Catalog No. 5000437428
Encompass_Preferred
Thalidomide-O-amido-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, and can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand and a linker
  • Used in the synthesis of PROTACs
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-004-37428 Supplier Medchemexpress LLC Supplier No. HY112618B100MG
Thalidomide-O-amido-C6-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker, and can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand and a linker
  • Used in the synthesis of PROTACs
  • Appearance: Solid
  • Color: White to off-white