Learn More
Medchemexpress LLC Thalidomide-O-amido-C6-NH2 (TFA) | 1950635-14-9 | 99.8% | 544.48 | 50 MG
Supplier: Medchemexpress LLC HY112618A50MG
Thalidomide-O-amido-C6-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the cereblon ligand and a linker. This compound is suitable for use in the synthesis of PROTACs, which function by exploiting the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
- E3 ligase ligand-linker conjugate
- Incorporates cereblon ligand and a linker
- Used in the synthesis of PROTACs
- PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins