Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-C8-NH2 (TFA) | 1950635-16-1 | MFCD31560485 | >=95.0% | 572.53 g/mol | C25H31F3N4O8 | 25 MG
SDP

Catalog No. 5000244608
Encompass_Preferred

Thalidomide-O-amido-C8-NH2 (TFA) is a thalidomide-derived cereblon ligand-linker conjugate supplied as the trifluoroacetic acid salt. It is intended for use as a building block in PROTAC synthesis and other E3 ligase ligand-linker research in chemical biology.

  • Chemical form: TFA salt.
  • Chemical formula: C25H31F3N4O8.
  • CAS number: 1950635-16-1.
  • Molecular weight: 572.53 g/mol.
  • Appearance: solid.
  • Typical use: building block for PROTAC and E3 ligase ligand-linker conjugate synthesis.
  • Purity: high (≥95%).

Catalog No. 50-002-44608 Supplier Medchemexpress LLC Supplier No. HY10361425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-C8-NH2 (TFA) is a thalidomide-derived cereblon ligand-linker conjugate supplied as the trifluoroacetic acid salt. It is intended for use as a building block in PROTAC synthesis and other E3 ligase ligand-linker research in chemical biology.

  • Chemical form: TFA salt.
  • Chemical formula: C25H31F3N4O8.
  • CAS number: 1950635-16-1.
  • Molecular weight: 572.53 g/mol.
  • Appearance: solid.
  • Typical use: building block for PROTAC and E3 ligase ligand-linker conjugate synthesis.
  • Purity: high (≥95%).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.