Learn More
Medchemexpress LLC Thalidomide-O-amido-C8-NHBoc | 1950635-34-3 | >98.0% | 558.6 g/mol | C28H38N4O8 | 10 MG
SDP
Catalog No. 5000298337
Encompass_Preferred
Thalidomide-O-amido-C8-NHBoc is a thalidomide-derived ligand-linker conjugate containing a thalidomide moiety, an amide linkage, an eight-carbon alkyl chain, and a Boc-protected amine (carbamate). It is supplied as a solid research reagent intended for use as a cereblon (E3 ligase) ligand building block in the synthesis of PROTACs and related medicinal chemistry applications.
  • Contains thalidomide core with an amide-linked C8 spacer.
  • Includes a Boc-protected terminal amine for selective deprotection and coupling.
  • High reported purity suitable for research applications.
  • Solid form with typical storage recommendations for small-molecule building blocks.
  • Used as a cereblon ligand-linker conjugate in targeted protein degradation research.
Catalog No. 50-002-98337 Supplier Medchemexpress LLC Supplier No. HY4278010MG
Thalidomide-O-amido-C8-NHBoc is a thalidomide-derived ligand-linker conjugate containing a thalidomide moiety, an amide linkage, an eight-carbon alkyl chain, and a Boc-protected amine (carbamate). It is supplied as a solid research reagent intended for use as a cereblon (E3 ligase) ligand building block in the synthesis of PROTACs and related medicinal chemistry applications.
  • Contains thalidomide core with an amide-linked C8 spacer.
  • Includes a Boc-protected terminal amine for selective deprotection and coupling.
  • High reported purity suitable for research applications.
  • Solid form with typical storage recommendations for small-molecule building blocks.
  • Used as a cereblon ligand-linker conjugate in targeted protein degradation research.