Learn More
Medchemexpress LLC Thalidomide-O-amido-C8-NHBoc | 1950635-34-3 | 98.4% | 558.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY427805MG
Encompass_Preferred
Thalidomide-O-amido-C8-NHBoc contains a Thalidomide group, an amide, an alkylC8 chain, and a carbamate protecting group.
Catalog No. 50-004-04651