Learn More
Medchemexpress LLC Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc | 1799711-31-1 | 98.4% | 634.67 | C30H42N4O11 | 5 MG
SDP
Catalog No. 5000221664
Encompass_Preferred
Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate containing a PEG3 spacer and a Boc-protected terminal amine. Supplied as a white to off-white solid at high purity, it is designed for use as a building block in PROTAC synthesis and targeted protein degradation research, enabling modular assembly of cereblon-recruiting degraders.
  • Thalidomide-based cereblon ligand useful for recruiting E3 ligase.
  • Peg3 linker provides a flexible spacer for conjugation to target ligands.
  • Boc-protected terminal amine enables orthogonal deprotection and coupling chemistry.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Available in small-mass pack sizes for discovery and early-stage research.
Catalog No. 50-002-21664 Supplier Medchemexpress LLC Supplier No. HY1451775MG
Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate containing a PEG3 spacer and a Boc-protected terminal amine. Supplied as a white to off-white solid at high purity, it is designed for use as a building block in PROTAC synthesis and targeted protein degradation research, enabling modular assembly of cereblon-recruiting degraders.
  • Thalidomide-based cereblon ligand useful for recruiting E3 ligase.
  • Peg3 linker provides a flexible spacer for conjugation to target ligands.
  • Boc-protected terminal amine enables orthogonal deprotection and coupling chemistry.
  • High purity suitable for synthetic and medicinal chemistry applications.
  • Available in small-mass pack sizes for discovery and early-stage research.