Learn More
Medchemexpress LLC Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc | 1799711-31-1 | 98.4% | C30H42N4O11 | 10MG
SDP
Catalog No. 5000260322
Encompass_Preferred
Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc is a thalidomide-derived E3 ligase ligand-linker conjugate supplied as a solid for research use. The compound features a PEG3 linker and a Boc-protected terminal amine, offered with high reported purity and characterized molecular data, suitable as a synthetic intermediate in chemical biology and PROTAC development.
  • High reported purity suitable for synthesis and analysis.
  • Characterized molecular formula and molecular weight for experimental planning.
  • Boc-protected terminal amine enables selective deprotection and conjugation.
  • PEG3 linker provides flexibility and improved solubility in conjugates.
  • Available in small milligram quantities for research workflows.
Catalog No. 50-002-60322 Supplier Medchemexpress LLC Supplier No. HY14517710MG
Thalidomide-O-amido-CH2-PEG3-CH2-NH-Boc is a thalidomide-derived E3 ligase ligand-linker conjugate supplied as a solid for research use. The compound features a PEG3 linker and a Boc-protected terminal amine, offered with high reported purity and characterized molecular data, suitable as a synthetic intermediate in chemical biology and PROTAC development.
  • High reported purity suitable for synthesis and analysis.
  • Characterized molecular formula and molecular weight for experimental planning.
  • Boc-protected terminal amine enables selective deprotection and conjugation.
  • PEG3 linker provides flexibility and improved solubility in conjugates.
  • Available in small milligram quantities for research workflows.