Learn More
Medchemexpress LLC Thalidomide-o-amido-PEG-C2-NH2 hydrochloride | 2204226-02-6 | 98.1% | 25 MG
SDP
Catalog No. 5000230957
Encompass_Preferred
Thalidomide-O-amido-PEG-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate provided as the hydrochloride salt for research use in PROTAC design and chemical biology applications.
  • Thalidomide-derived CRBN ligand-linker conjugate for recruiting cereblon.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity suitable for research (98.1%).
  • Designed for conjugation and PROTAC synthesis workflows.
  • Small 25 mg quantity appropriate for exploratory experiments.
Catalog No. 50-002-30957 Supplier Medchemexpress LLC Supplier No. HY122694A25MG
Thalidomide-O-amido-PEG-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) ligand-linker conjugate provided as the hydrochloride salt for research use in PROTAC design and chemical biology applications.
  • Thalidomide-derived CRBN ligand-linker conjugate for recruiting cereblon.
  • Hydrochloride salt form for improved solubility and handling.
  • High purity suitable for research (98.1%).
  • Designed for conjugation and PROTAC synthesis workflows.
  • Small 25 mg quantity appropriate for exploratory experiments.