Learn More
Medchemexpress LLC HY-P7024 10ug Medchemexpress, IFN-beta (HEK293-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970555
Encompass_Preferred
Medchemexpress, HY-112600A 500mg Thalidomide-O-amido-PEG1-(C1-​PEG)2-C2-NH2 CAS: Thalidomide-O-amido-PEG1-(C1-​PEG)2-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0555 Supplier Medchemexpress LLC Supplier No. HYP702410UG
Medchemexpress, HY-112600A 500mg Thalidomide-O-amido-PEG1-(C1-​PEG)2-C2-NH2 CAS: Thalidomide-O-amido-PEG1-(C1-​PEG)2-C2-NH2 is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.