Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG1-(C1-PEG)2-C2-NH2 TFA | 2435572-49-7 | 99.28% | 648.58 | 50 MG
SDP
Catalog No. 5000437962
Encompass_Preferred
Thalidomide-O-amido-PEG1-(C1-PEG)2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. At equivalent molar concentrations, both the salt and free forms of a compound exhibit comparable biological activity. Nevertheless, the salt form usually boasts enhanced water solubility and stability.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Features a 3-unit PEG linker for PROTAC technology
  • Salt form offers enhanced water solubility and stability
  • For research use only
Catalog No. 50-004-37962 Supplier Medchemexpress LLC Supplier No. HY112600A50MG
Thalidomide-O-amido-PEG1-(C1-PEG)2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 3-unit PEG linker used in PROTAC technology. At equivalent molar concentrations, both the salt and free forms of a compound exhibit comparable biological activity. Nevertheless, the salt form usually boasts enhanced water solubility and stability.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Features a 3-unit PEG linker for PROTAC technology
  • Salt form offers enhanced water solubility and stability
  • For research use only