Learn More
Medchemexpress LLC HY-P7043A 10ug Medchemexpress, IL-5 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501970559
Encompass_Preferred
Medchemexpress, HY-112617A 100mg Thalidomide-O-amido-PEG2-C2-NH2 (TFA) CAS:1957235-75-4 Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0559 Supplier Medchemexpress LLC Supplier No. HYP7043A10UG
Medchemexpress, HY-112617A 100mg Thalidomide-O-amido-PEG2-C2-NH2 (TFA) CAS:1957235-75-4 Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.