Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG2-C2-NH2 hydrochloride | 2376990-30-4 | 498.91 | 100 MG
SDP
Catalog No. 5000437275
Encompass_Preferred
Thalidomide-O-amido-PEG2-C2-NH2 hydrochloride incorporates an E3 ligase ligand and a linker, and can be an immunomodulator for the treatment of cancer. It is for research use only and not sold to patients. PROTACs contain two different ligands connected by a linker, one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates an E3 ligase ligand and a linker
  • Can be an immunomodulator for the treatment of cancer
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-004-37275 Supplier Medchemexpress LLC Supplier No. HY112617B100MG
Thalidomide-O-amido-PEG2-C2-NH2 hydrochloride incorporates an E3 ligase ligand and a linker, and can be an immunomodulator for the treatment of cancer. It is for research use only and not sold to patients. PROTACs contain two different ligands connected by a linker, one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Incorporates an E3 ligase ligand and a linker
  • Can be an immunomodulator for the treatment of cancer
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins