Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG2-C2-NH2 hydrochloride | 2376990-30-4 | 498.91 | 25 MG
SDP
Catalog No. 5000438844
Encompass_Preferred
Thalidomide-O-amido-PEG2-C2-NH2 hydrochloride incorporates an E3 ligase ligand and a linker, and can act as an immunomodulator for the treatment of cancer. It is for research use only and not sold to patients.
  • PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-004-38844 Supplier Medchemexpress LLC Supplier No. 25MGHY112617B
Thalidomide-O-amido-PEG2-C2-NH2 hydrochloride incorporates an E3 ligase ligand and a linker, and can act as an immunomodulator for the treatment of cancer. It is for research use only and not sold to patients.
  • PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.