Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG2-C2-NH2 (TFA) | 1957235-75-4 | 576.48 | 50 MG
SDP
Catalog No. 5000437426
Encompass_Preferred
Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide-based cereblon ligand and a 2-unit PEG linker used in PROTAC technology. This product is intended for research use only and for experienced personnel in appropriately equipped facilities.
  • Incorporates Thalidomide-based cereblon ligand
  • Features a 2-unit PEG linker
  • Used in PROTAC technology
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Identified as laboratory chemicals for substance manufacture
Catalog No. 50-004-37426 Supplier Medchemexpress LLC Supplier No. HY112617A50MG
Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide-based cereblon ligand and a 2-unit PEG linker used in PROTAC technology. This product is intended for research use only and for experienced personnel in appropriately equipped facilities.
  • Incorporates Thalidomide-based cereblon ligand
  • Features a 2-unit PEG linker
  • Used in PROTAC technology
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Identified as laboratory chemicals for substance manufacture