Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG2-C2-NH2 (TFA) | 1957235-75-4 | 576.48 | 500 MG
SDP
Catalog No. 5000436230
Encompass_Preferred
Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a 2-unit PEG linker used in PROTAC technology. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Uses a 2-unit PEG linker
  • Utilized in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-004-36230 Supplier Medchemexpress LLC Supplier No. HY112617A500MG
Thalidomide-O-amido-PEG2-C2-NH2 TFA is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a 2-unit PEG linker used in PROTAC technology. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Uses a 2-unit PEG linker
  • Utilized in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins