Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG3-C2-NH2 hydrochloride | 2245697-84-9 | MFCD31656717 | 98.3% | 542.97 | C23H31ClN4O9 | 250 MG
SDP
Catalog No. 5000241049
Encompass_Preferred
Thalidomide-O-amido-PEG3-C2-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate used as an intermediate in the design and synthesis of PROTACs and targeted protein degraders. Supplied as the hydrochloride salt, it is provided as an off-white to yellow solid with high reported purity for research applications.
  • High purity suitable for research and synthetic use.
  • Hydrochloride salt form for improved handling and stability.
  • Good solubility in DMSO and water; may require ultrasonic aid.
  • Available in multiple package sizes, including 250 mg.
  • Recommended storage at -20°C sealed, away from moisture.
Catalog No. 50-002-41049 Supplier Medchemexpress LLC Supplier No. HY107440A250MG
Thalidomide-O-amido-PEG3-C2-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate used as an intermediate in the design and synthesis of PROTACs and targeted protein degraders. Supplied as the hydrochloride salt, it is provided as an off-white to yellow solid with high reported purity for research applications.
  • High purity suitable for research and synthetic use.
  • Hydrochloride salt form for improved handling and stability.
  • Good solubility in DMSO and water; may require ultrasonic aid.
  • Available in multiple package sizes, including 250 mg.
  • Recommended storage at -20°C sealed, away from moisture.