Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG3-C2-NH2 hydrochloride | 2245697-84-9 | MFCD31656717 | 98.3% | 542.97 | C23H31ClN4O9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY107440A50MG
Encompass_Preferred
Thalidomide-O-amido-PEG3-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ligase ligand provided as a hydrochloride salt for research use in PROTAC design and chemical biology. Supplied as a solid with defined molecular properties and high reported purity, it serves as a linker/ligand building block for medicinal chemistry and degradation probe synthesis.
  • High purity (>98%).
  • Solid, off-white to yellow appearance.
  • Molecular formula C23H31ClN4O9; molecular weight 542.97.
  • Suitable as a CRBN E3 ligase ligand in PROTAC and protein degradation studies.
  • Available in milligram-scale quantities for laboratory research.
Catalog No. 50-002-40460