Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-PEG3-C2-NH2 TFA | 1957236-21-3 | 99.6% | 620.53 g/mol | C25H31F3N4O11 | 10 MG
SDP

Catalog No. 5000247673
Encompass_Preferred

Thalidomide-O-amido-PEG3-C2-NH2 TFA is a thalidomide-based cereblon ligand-linker conjugate (trifluoroacetic acid salt) designed as a building block for PROTAC synthesis. It links a thalidomide-derived E3 ligase ligand to a three-unit PEG spacer terminating in a C2 amino handle, enabling attachment to targeting ligands and facilitating cereblon-mediated target degradation.

  • Thalidomide-based cereblon ligand for E3 ligase recruitment.
  • Three-unit PEG linker providing flexible spacing and solubility.
  • C2 amino terminal suitable for amide or urea coupling.
  • Trifluoroacetic acid salt form for improved handling and stability.
  • Molecular formula C25H31F3N4O11; molecular weight 620.53 g/mol.
  • Soluble in DMSO at 50 mg/mL; ultrasonic may be required.
  • Recommended storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities suitable for synthesis, e.g., 10 mg.

Catalog No. 50-002-47673 Supplier Medchemexpress LLC Supplier No. HY10361110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-PEG3-C2-NH2 TFA is a thalidomide-based cereblon ligand-linker conjugate (trifluoroacetic acid salt) designed as a building block for PROTAC synthesis. It links a thalidomide-derived E3 ligase ligand to a three-unit PEG spacer terminating in a C2 amino handle, enabling attachment to targeting ligands and facilitating cereblon-mediated target degradation.

  • Thalidomide-based cereblon ligand for E3 ligase recruitment.
  • Three-unit PEG linker providing flexible spacing and solubility.
  • C2 amino terminal suitable for amide or urea coupling.
  • Trifluoroacetic acid salt form for improved handling and stability.
  • Molecular formula C25H31F3N4O11; molecular weight 620.53 g/mol.
  • Soluble in DMSO at 50 mg/mL; ultrasonic may be required.
  • Recommended storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities suitable for synthesis, e.g., 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.