Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG3-C2-NH2 TFA | 1957236-21-3 | MFCD31560482 | >98.0% | 572.5 g/mol | C24H27F3N4O9 | 5 MG
SDP
Catalog No. 5000244984
Encompass_Preferred
Thalidomide-O-amido-PEG3-C2-NH2 TFA is a thalidomide-based cereblon E3 ligase ligand-linker conjugate provided as the trifluoroacetic acid (TFA) salt. It is intended as a building block for PROTAC synthesis, combining a thalidomide-derived CRBN-binding moiety with a three-unit polyethylene glycol (PEG3) spacer and a short C2 amine handle for conjugation.
  • Provides a cereblon-binding thalidomide ligand for PROTAC design.
  • Includes a PEG3 linker to improve solubility and flexibility.
  • Features a C2 amine handle for facile coupling to warheads.
  • Offered as the TFA salt to aid handling and stability.
  • Supplied at high purity (>98%) suitable for synthetic use.
  • Available in small research-scale quantities for method development.
Catalog No. 50-002-44984 Supplier Medchemexpress LLC Supplier No. HY1036115MG
Thalidomide-O-amido-PEG3-C2-NH2 TFA is a thalidomide-based cereblon E3 ligase ligand-linker conjugate provided as the trifluoroacetic acid (TFA) salt. It is intended as a building block for PROTAC synthesis, combining a thalidomide-derived CRBN-binding moiety with a three-unit polyethylene glycol (PEG3) spacer and a short C2 amine handle for conjugation.
  • Provides a cereblon-binding thalidomide ligand for PROTAC design.
  • Includes a PEG3 linker to improve solubility and flexibility.
  • Features a C2 amine handle for facile coupling to warheads.
  • Offered as the TFA salt to aid handling and stability.
  • Supplied at high purity (>98%) suitable for synthetic use.
  • Available in small research-scale quantities for method development.