Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-PEG3-C2-NH2 TFA | 1957236-21-3 | MFCD31560482 | 99.6% | 660.67 g/mol | C23H30F3N5O9 | 25 MG
SDP

Catalog No. 5000244963
Encompass_Preferred

Thalidomide-O-amido-PEG3-C2-NH2 TFA is a trifluoroacetate salt E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a three-unit polyethylene glycol (PEG3) linker for use in PROTAC synthesis and related chemical biology research. It is supplied as a high-purity solid and is intended for research use only.

  • Contains a thalidomide-based cereblon ligand for cereblon-mediated degradation studies.
  • PEG3 linker provides flexibility and solubility for linker optimization in PROTAC design.
  • Offered as a solid TFA salt for convenient handling and storage.
  • High purity suitable for synthetic chemistry and biological probe development.
  • Recommended storage: powder at -20°C; in solution, store at -80°C for long-term preservation.

Catalog No. 50-002-44963 Supplier Medchemexpress LLC Supplier No. HY10361125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-PEG3-C2-NH2 TFA is a trifluoroacetate salt E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a three-unit polyethylene glycol (PEG3) linker for use in PROTAC synthesis and related chemical biology research. It is supplied as a high-purity solid and is intended for research use only.

  • Contains a thalidomide-based cereblon ligand for cereblon-mediated degradation studies.
  • PEG3 linker provides flexibility and solubility for linker optimization in PROTAC design.
  • Offered as a solid TFA salt for convenient handling and storage.
  • High purity suitable for synthetic chemistry and biological probe development.
  • Recommended storage: powder at -20°C; in solution, store at -80°C for long-term preservation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.