Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG4-azide | 2411681-89-3 | 99.2% | C25H32N6O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14101150MG
Encompass_Preferred
Thalidomide-O-amido-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-55057