Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride | 2245697-85-0 | 95.0% | 100 MG
SDP
Catalog No. 5000231014
Encompass_Preferred
Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-derived cereblon ligand linked to a PEG4-C2 amine linker; supplied as the hydrochloride salt and intended as a building block for PROTAC and E3 ligase ligand-linker conjugates in research applications.
  • Functions as an E3 ligase ligand-linker conjugate for PROTAC synthesis.
  • Contains a thalidomide-derived cereblon ligand for target engagement.
  • Includes a PEG4-based linker to provide flexibility and solubility.
  • Supplied as a hydrochloride salt for improved handling.
  • Typical purity ≥95% suitable for research use.
Catalog No. 50-002-31014 Supplier Medchemexpress LLC Supplier No. HY122710A100MG
Add to Cart
Add to Cart
Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-derived cereblon ligand linked to a PEG4-C2 amine linker; supplied as the hydrochloride salt and intended as a building block for PROTAC and E3 ligase ligand-linker conjugates in research applications.
  • Functions as an E3 ligase ligand-linker conjugate for PROTAC synthesis.
  • Contains a thalidomide-derived cereblon ligand for target engagement.
  • Includes a PEG4-based linker to provide flexibility and solubility.
  • Supplied as a hydrochloride salt for improved handling.
  • Typical purity ≥95% suitable for research use.