Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride | 2245697-85-0 | 95.0% | 25 MG
SDP

Catalog No. 5000231015
Encompass_Preferred

Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate supplied as the hydrochloride salt. It is intended for use as a building block in PROTAC design and chemical biology studies to recruit the cereblon E3 ligase.

  • Hydrochloride salt for improved stability and handling.
  • Contains a PEG4 linker with a terminal C2 amine for facile conjugation.
  • Suitable as an E3 ligase ligand in PROTAC design and CRBN recruitment studies.
  • Supplied in small research quantities for medicinal chemistry applications.
  • Characterized by molecular weight 587.02 and formula C25H35ClN4O10.

Catalog No. 50-002-31015 Supplier Medchemexpress LLC Supplier No. HY122710A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate supplied as the hydrochloride salt. It is intended for use as a building block in PROTAC design and chemical biology studies to recruit the cereblon E3 ligase.

  • Hydrochloride salt for improved stability and handling.
  • Contains a PEG4 linker with a terminal C2 amine for facile conjugation.
  • Suitable as an E3 ligase ligand in PROTAC design and CRBN recruitment studies.
  • Supplied in small research quantities for medicinal chemistry applications.
  • Characterized by molecular weight 587.02 and formula C25H35ClN4O10.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.