Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride | 2245697-85-0 | 95.0% | 50 MG
SDP

Catalog No. 5000231016
Encompass_Preferred

Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for research use in targeted protein degradation and linker chemistry. It has a reported purity of 95.0%, molecular weight 587.02, and formula C25H35ClN4O10, and is offered in small research sizes including 50 MG.

  • Thalidomide-based cereblon ligand-linker conjugate for targeted protein degradation research.
  • Hydrochloride salt form for research handling.
  • Reported purity 95.0% suitable for research applications.
  • Molecular weight 587.02 and chemical formula C25H35ClN4O10.
  • Available in small research sizes including 50 MG.

Catalog No. 50-002-31016 Supplier Medchemexpress LLC Supplier No. HY122710A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-amido-PEG4-C2-NH2 hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ligase ligand-linker conjugate supplied as the hydrochloride salt for research use in targeted protein degradation and linker chemistry. It has a reported purity of 95.0%, molecular weight 587.02, and formula C25H35ClN4O10, and is offered in small research sizes including 50 MG.

  • Thalidomide-based cereblon ligand-linker conjugate for targeted protein degradation research.
  • Hydrochloride salt form for research handling.
  • Reported purity 95.0% suitable for research applications.
  • Molecular weight 587.02 and chemical formula C25H35ClN4O10.
  • Available in small research sizes including 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.