Learn More
Medchemexpress LLC Thalidomide-O-amido-PEG4-propargyl | 2496687-00-2 | 98.0% | C26H31N3O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14101350MG
Encompass_Preferred
Thalidomide-O-amido-PEG4-propargyl is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54974