Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-C2-acid | 2369068-42-6 | 95.0% | 346.29 g/mol | C16H14N2O7 | 5 MG
SDP

Catalog No. 5000262675
Encompass_Preferred

Thalidomide-O-C2-acid is a synthesized E3 ligase ligand-linker conjugate that incorporates a thalidomide-based cereblon ligand with a two-carbon linker. It is intended as a research-grade building block for PROTAC synthesis and related chemical biology applications, and is supplied with reported analytical data suitable for experimental use.

  • E3 ligase ligand-linker conjugate for PROTAC design.
  • Molecular formula C16H14N2O7.
  • Molecular weight 346.29 g/mol.
  • CAS number 2369068-42-6.
  • Reported purity 95.0%.
  • Available in small milligram pack sizes for research use.

Catalog No. 50-002-62675 Supplier Medchemexpress LLC Supplier No. HY1318805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-C2-acid is a synthesized E3 ligase ligand-linker conjugate that incorporates a thalidomide-based cereblon ligand with a two-carbon linker. It is intended as a research-grade building block for PROTAC synthesis and related chemical biology applications, and is supplied with reported analytical data suitable for experimental use.

  • E3 ligase ligand-linker conjugate for PROTAC design.
  • Molecular formula C16H14N2O7.
  • Molecular weight 346.29 g/mol.
  • CAS number 2369068-42-6.
  • Reported purity 95.0%.
  • Available in small milligram pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.