Learn More
Medchemexpress LLC Thalidomide-O-C2-acid | 2369068-42-6 | 95.0% | C16H14N2O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13188010MG
Encompass_Preferred
Thalidomide-O-C2-acid is an E3 ligase ligand-linker conjugate that pairs a thalidomide-derived cereblon ligand with a two-carbon linker. It is designed as a research building block for proteolysis targeting chimera (PROTAC) construction and targeted protein degradation studies, and is supplied with analytical data supporting its identity and purity.
  • Contains a thalidomide-derived cereblon ligand for CRBN recruitment.
  • Includes a short two-carbon linker suitable for conjugation.
  • Provided with reported purity and molecular characterization.
  • Available in multiple small pack sizes for research-scale synthesis.
  • Intended for use as a building block in PROTAC synthesis and chemical biology studies.
  • Supplied with an accessible datasheet and safety documentation.
Catalog No. 50-002-60589