Learn More
Medchemexpress LLC Thalidomide-O-C4-COOH | 2169266-67-3 | 97.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1309501G
Encompass_Preferred
Thalidomide-O-C4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is for research use only and not intended for sale to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40580