Learn More
Medchemexpress LLC Thalidomide-O-C4-COOH | 2169266-67-3 | 97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130950100MG
Encompass_Preferred
Thalidomide-O-C4-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is for research use only and not sold to patients.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • For research use only
Catalog No. 50-003-41007