Learn More
Medchemexpress LLC Thalidomide-O-C4-COOH | 2169266-67-3 | 97.0% | 50 MG
SDP
Catalog No. 5000338553
Encompass_Preferred
Thalidomide-O-C4-COOH is a synthesized E3 ligase ligand-linker conjugate designed for research purposes. It integrates a thalidomide-based cereblon ligand and a linker, both crucial components utilized in PROTAC (Proteolysis Targeting Chimeras) technology. This product is not intended for patient use.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a thalidomide-based cereblon ligand
  • Includes a linker commonly used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Appears as a white to off-white solid
  • Available in various quantities with specific storage guidelines
  • Supplied with a Data Sheet, Certificate of Analysis (COA), Safety Data Sheet (SDS), and Handling Instructions
Catalog No. 50-003-38553 Supplier Medchemexpress LLC Supplier No. HY13095050MG
Thalidomide-O-C4-COOH is a synthesized E3 ligase ligand-linker conjugate designed for research purposes. It integrates a thalidomide-based cereblon ligand and a linker, both crucial components utilized in PROTAC (Proteolysis Targeting Chimeras) technology. This product is not intended for patient use.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a thalidomide-based cereblon ligand
  • Includes a linker commonly used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Appears as a white to off-white solid
  • Available in various quantities with specific storage guidelines
  • Supplied with a Data Sheet, Certificate of Analysis (COA), Safety Data Sheet (SDS), and Handling Instructions