Learn More
Medchemexpress LLC Thalidomide-O-C6-azide | 2411389-65-4 | 99.1% | 399.40 | 100 MG
SDP
Catalog No. 5000360939
Encompass_Preferred
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate, incorporating the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This click chemistry reagent contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. It is designed for research applications related to PROTACs, which utilize the ubiquitin-proteasome system to degrade target proteins.
  • Incorporates cereblon ligand and a linker for PROTAC technology
  • Click chemistry reagent with an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Designed for research in protein degradation via PROTACs
Catalog No. 50-003-60939 Supplier Medchemexpress LLC Supplier No. HY148556100MG
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate, incorporating the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This click chemistry reagent contains an Azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. It is designed for research applications related to PROTACs, which utilize the ubiquitin-proteasome system to degrade target proteins.
  • Incorporates cereblon ligand and a linker for PROTAC technology
  • Click chemistry reagent with an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Designed for research in protein degradation via PROTACs