Learn More
Medchemexpress LLC Thalidomide-O-C6-azide | 2411389-65-4 | 99.1% | 399.40 | 50 MG
SDP
Catalog No. 5000360598
Encompass_Preferred
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates a thalidomide-based cereblon ligand and a linker used in PROTAC technology. This product is also a click chemistry reagent, containing an azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a thalidomide-based cereblon ligand
  • Features a linker used in PROTAC technology
  • Functions as a click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Allows strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Supports selective degradation of target proteins through PROTACs
Catalog No. 50-003-60598 Supplier Medchemexpress LLC Supplier No. HY14855650MG
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates a thalidomide-based cereblon ligand and a linker used in PROTAC technology. This product is also a click chemistry reagent, containing an azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a thalidomide-based cereblon ligand
  • Features a linker used in PROTAC technology
  • Functions as a click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Allows strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Supports selective degradation of target proteins through PROTACs