Learn More
Medchemexpress LLC Thalidomide-O-C6-azide | 2411389-65-4 | 99.1% | 399.40 g·mol⁻¹ | C19H21N5O5 | 10 MG
SDP
Catalog No. 5000287144
Encompass_Preferred
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a six-carbon azide-containing linker. It is intended as a building block for PROTAC design and related targeted protein degradation research, providing a clickable azide handle for conjugation or labeling in chemical biology applications.
  • High purity suitable for research applications (≈99.1%).
  • Azide functional group enables copper-free and copper-catalyzed click chemistry.
  • Six-carbon linker provides flexible spacing for bifunctional molecule design.
  • Storage-stable when sealed at low temperature, with extended stability in solvent at -80°C.
  • Available in multiple small-scale quantities for synthesis and assay development.
Catalog No. 50-002-87144 Supplier Medchemexpress LLC Supplier No. HY14855610MG
Thalidomide-O-C6-azide is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a six-carbon azide-containing linker. It is intended as a building block for PROTAC design and related targeted protein degradation research, providing a clickable azide handle for conjugation or labeling in chemical biology applications.
  • High purity suitable for research applications (≈99.1%).
  • Azide functional group enables copper-free and copper-catalyzed click chemistry.
  • Six-carbon linker provides flexible spacing for bifunctional molecule design.
  • Storage-stable when sealed at low temperature, with extended stability in solvent at -80°C.
  • Available in multiple small-scale quantities for synthesis and assay development.