Learn More
Medchemexpress LLC Thalidomide-O-C6-COOH | 2169266-69-5 | 99.29% | 50 MG
SDP
Catalog No. 5000340577
Encompass_Preferred
Thalidomide-O-C6-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, which is used in PROTAC (Proteolysis-Targeting Chimera) technology. This compound is intended for research use only.
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Composed of two different ligands connected by a linker
Catalog No. 50-003-40577 Supplier Medchemexpress LLC Supplier No. HY13095150MG
Thalidomide-O-C6-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, which is used in PROTAC (Proteolysis-Targeting Chimera) technology. This compound is intended for research use only.
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Composed of two different ligands connected by a linker