Learn More
Medchemexpress LLC Thalidomide-O-C6-COOH | 2169266-69-5 | 99.3% | 100 MG
SDP
Catalog No. 5000340309
Encompass_Preferred
Thalidomide-O-C6-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, designed for use in PROTAC technology. PROTACs function by connecting two ligands: one for an E3 ubiquitin ligase and another for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Designed for PROTAC technology applications
  • Selectively degrades target proteins via ubiquitin-proteasome system
  • Off-white to light yellow solid
  • Soluble in DMSO at ≥ 100 mg/mL
Catalog No. 50-003-40309 Supplier Medchemexpress LLC Supplier No. HY130951100MG
Thalidomide-O-C6-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, designed for use in PROTAC technology. PROTACs function by connecting two ligands: one for an E3 ubiquitin ligase and another for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Designed for PROTAC technology applications
  • Selectively degrades target proteins via ubiquitin-proteasome system
  • Off-white to light yellow solid
  • Soluble in DMSO at ≥ 100 mg/mL