Learn More
Medchemexpress LLC Thalidomide-O-C6-NHBoc | 2509093-23-4 | 96.0% | C24H31N3O7 | 25 MG
SDP
Catalog No. 5000352622
Encompass_Preferred
Thalidomide-O-C6-NHBoc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Targets E3 ligase ligand-linker conjugates, apoptosis, and autophagy
  • Targets cereblon
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-52622 Supplier Medchemexpress LLC Supplier No. 25MGHY141887
Thalidomide-O-C6-NHBoc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Targets E3 ligase ligand-linker conjugates, apoptosis, and autophagy
  • Targets cereblon
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 100 mg/mL