Learn More
Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | 95.1% | 1 ML
SDP
Catalog No. 5000338832
Encompass_Preferred
Thalidomide-O-C8-Boc is a Thalidomide-based Cereblon ligand utilized for the recruitment of CRBN protein. It can be linked to a target protein ligand via a linker to create PROTACs.
  • Cereblon ligand
  • Used for recruitment of CRBN protein
  • Can be linked to a target protein ligand via a linker to create PROTACs
  • For research use only
Catalog No. 50-003-38832 Supplier Medchemexpress LLC Supplier No. HY13115910MM/1ML
Thalidomide-O-C8-Boc is a Thalidomide-based Cereblon ligand utilized for the recruitment of CRBN protein. It can be linked to a target protein ligand via a linker to create PROTACs.
  • Cereblon ligand
  • Used for recruitment of CRBN protein
  • Can be linked to a target protein ligand via a linker to create PROTACs
  • For research use only