Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | 95.1% | 100 MG
SDP

Catalog No. 5000338626
Encompass_Preferred

Thalidomide-O-C8-Boc is a Thalidomide-based Cereblon ligand designed for recruiting CRBN protein. It can be linked to a target protein ligand via a linker to form PROTACs, making it suitable for research use in laboratory chemical manufacturing. This substance is stable under recommended storage conditions and is not classified as a hazardous substance or mixture.

  • Used to recruit CRBN protein
  • Can be linked to a target protein ligand via a linker to form PROTACs
  • Suitable for laboratory chemical manufacturing
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture
  • Recommended storage temperature: -20°C, stored under nitrogen
  • Molecular formula: C26H34N2O7
  • Molecular weight: 486.56
  • Appearance: Solid, white to off-white

Catalog No. 50-003-38626 Supplier Medchemexpress LLC Supplier No. HY131159100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-C8-Boc is a Thalidomide-based Cereblon ligand designed for recruiting CRBN protein. It can be linked to a target protein ligand via a linker to form PROTACs, making it suitable for research use in laboratory chemical manufacturing. This substance is stable under recommended storage conditions and is not classified as a hazardous substance or mixture.

  • Used to recruit CRBN protein
  • Can be linked to a target protein ligand via a linker to form PROTACs
  • Suitable for laboratory chemical manufacturing
  • Stable under recommended storage conditions
  • Not classified as a hazardous substance or mixture
  • Recommended storage temperature: -20°C, stored under nitrogen
  • Molecular formula: C26H34N2O7
  • Molecular weight: 486.56
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.