Learn More
Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | 95.1% | 25 MG
SDP
Catalog No. 5000339519
Encompass_Preferred
Thalidomide-O-C8-Boc is a thalidomide-based Cereblon ligand used for the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTACs.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be used to form PROTACs when connected to a ligand for protein by a linker
  • Appearance: Solid, white to off-white
  • For research use only
Catalog No. 50-003-39519 Supplier Medchemexpress LLC Supplier No. 25MGHY131159
Thalidomide-O-C8-Boc is a thalidomide-based Cereblon ligand used for the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTACs.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be used to form PROTACs when connected to a ligand for protein by a linker
  • Appearance: Solid, white to off-white
  • For research use only