Learn More
Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | 95.1% | 250 MG
SDP
Catalog No. 5000343019
Encompass_Preferred
Thalidomide-O-C8-Boc is a Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be linked to a ligand for protein via a linker to create PROTACs. This product is a white to off-white solid, stable under recommended storage conditions, and intended for research use only.
  • Cereblon ligand
  • Utilized in CRBN protein recruitment
  • Can be linked to a ligand for protein via a linker to create PROTACs
  • Solid appearance
  • White to off-white color
  • Stable under recommended storage conditions
  • Research use only
Catalog No. 50-003-43019 Supplier Medchemexpress LLC Supplier No. 250MGHY131159
Thalidomide-O-C8-Boc is a Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be linked to a ligand for protein via a linker to create PROTACs. This product is a white to off-white solid, stable under recommended storage conditions, and intended for research use only.
  • Cereblon ligand
  • Utilized in CRBN protein recruitment
  • Can be linked to a ligand for protein via a linker to create PROTACs
  • Solid appearance
  • White to off-white color
  • Stable under recommended storage conditions
  • Research use only